Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028048.2 | NP_082324.1 | 1 | 891 | ||
GenBank mRNA | AK007453.1 | - | 1 | 893 | ||
AK035739.1 | - | 874 | ||||
AK157873.1 | - | 1 | 885 | |||
AK171131.1 | - | 1 | 898 | |||
BC004569.1 | - | 1 | 331 | |||
BC019996.1 | - | 1 | 833 | 4207271 |