Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028250.3 | NP_082526.2 | 6 | 1118 | ||
GenBank mRNA | AA260937.1 | - | 1 | 21 | ||
AK011781.1 | - | 6 | 1111 | |||
AK167959.1 | - | 7 | 1058 | |||
BC115580.1 | - | 6 | 710 | 40102241 | ||
BC115581.1 | - | 6 | 710 | 40102242 |