Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028846.5 | NP_083122.1 | 17 | 1949 | ||
GenBank mRNA | AF449715.1 | - | 17 | 1900 | ||
AK029890.1 | - | 1845 | ||||
AK047242.1 | - | 17 | 1868 | |||
AK054279.1 | - | 17 | 1892 | |||
AK087412.1 | - | 17 | 1860 | |||
AK163663.1 | - | 17 | 1859 | |||
AK173083.1 | - | 17 | 1851 | |||
AK186822.1 | - | 1 | 25 | |||
BC019735.1 | - | 3 | 149 | |||
BC037199.1 | - | 11 | 1333 | |||
BC079674.1 | - | 1838 | 30531994 |