Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amotl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081395.1 | NP_001074864.1 | 12, 13 | 3021 | ||
GenBank mRNA | AK016526.1 | - | 13, 14 | 2881 | ||
AK220238.1 | - | 10, 11 | 2255 | |||
AX763684.1 | - | 10, 11 | 2545 | |||
BC055117.1 | - | 4, 5 | 746 | |||
BC151000.1 | - | 12, 13 | 2829 | 9087913 | ||
BC151004.1 | - | 12, 13 | 2829 | 9087917 | ||
BC151007.1 | - | 12, 13 | 2829 | 9087920 |