Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdyhv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029734.1 | NP_084010.1 | 5, 6 | 571 | ||
GenBank mRNA | AK150578.1 | - | 5, 6 | 623 | ||
BC051524.1 | - | 5, 6 | 571 | 1514917 |