Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mboat7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029934.3 | NP_084210.2 | 8 | 1985 | ||
GenBank mRNA | AB055410.1 | - | 8 | 1983 | ||
AK019981.1 | - | 8 | 1396 | |||
AK150010.1 | - | 8 | 1379 | |||
AK154606.1 | - | 8 | 1984 | |||
AK170124.1 | - | 8 | 1462 | |||
BC023417.1 | - | 8 | 1433 | 5028759 | ||
BC118958.2 | - | 8 | 1630 | 40093821 | ||
EU016380.1 | - | 7 | 1197 |