Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mir99ahg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021336.1 | - | 845 | |||
AK033222.1 | - | 406 | ||||
AK082961.1 | - | 906 |