Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gins3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030198.3 | NP_084474.1 | 3 | 499 | ||
GenBank mRNA | AK019225.1 | - | 3 | 500 | ||
AK146574.1 | - | 3 | 530 | |||
AK150602.1 | - | 3 | 531 | |||
AK151243.1 | - | 3 | 531 | |||
AK152572.1 | - | 3 | 531 | |||
AK160606.1 | - | 3 | 500 | |||
BC039794.1 | - | 3 | 496 | 5121680 | ||
BC064746.1 | - | 3 | 488 | 30095310 |