Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290688.1 | NP_001277617.1 | 4 | 321 | ||
NM_020000.3 | NP_064384.2 | 4 | 361 | |||
NM_173719.3 | NP_776067.1 | 5 | 409 | |||
GenBank mRNA | AB041805.1 | - | 3 | 1847 | ||
AK005422.1 | - | 5 | 392 | |||
AK008759.1 | - | 4 | 340 | |||
AK011080.1 | - | 4 | 321 | |||
AK147032.1 | - | 4 | 347 | |||
AK148025.1 | - | 4 | 342 | |||
BC051683.1 | - | 4 | 299 |