Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030709.2 | NP_109634.2 | 10, 11 | 1149 | ||
GenBank mRNA | AB016229.1 | - | 10, 11 | 1147 | ||
AB016230.1 | - | 10, 11 | 1149 | |||
AB016423.1 | - | 6, 7 | 764 | |||
AB041037.1 | - | 10, 11 | 1106 | |||
AK029714.1 | - | 11, 12 | 1996 | |||
BC132151.1 | - | 9, 10 | 1095 | 40130428 | ||
BC145027.1 | - | 9, 10 | 1077 | 9053575 |