Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pum2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160219.1 | NP_001153691.1 | 20 | 3159 | ||
NM_001160220.1 | NP_001153692.1 | 20 | 3153 | |||
NM_001160221.1 | NP_001153693.1 | 20 | 2968 | |||
NM_001160222.1 | NP_001153694.1 | 19 | 2731 | |||
NM_030723.2 | NP_109648.2 | 20 | 3083 | |||
GenBank mRNA | AF315590.1 | - | 20 | 3099 | ||
AK048119.1 | - | 14 | 2022 | |||
AK122225.1 | - | 21 | 3001 | |||
AK134185.1 | - | 20 | 2967 | |||
AK141682.1 | - | 20 | 3015 | |||
AK147617.1 | - | 21 | 3101 | |||
AK161357.1 | - | 9 | 1324 | |||
AK163964.1 | - | 19 | 2706 | |||
AY027917.1 | - | 20 | 2999 | |||
BC013765.1 | - | 4 | 334 | |||
BC041773.1 | - | 18 | 2705 | 3986207 |