Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252577.1 | NP_001239506.1 | 6 | 725 | ||
NM_010649.3 | NP_034779.1 | 6 | 728 | |||
GenBank mRNA | AF218078.1 | - | 5 | 637 | ||
AF218079.1 | - | 5 | 637 | |||
AK080171.1 | - | 6 | 726 | |||
AK145141.1 | - | 5 | 596 | |||
AY860973.1 | - | 580 | ||||
AY860975.1 | - | 5 | 499 | |||
L78247.1 | - | 5 | 580 | |||
U10090.1 | - | 5 | 638 |
Gene Symbol: Klra12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010646.1 | NP_034776.1 | 5 | 643 | ||
GenBank mRNA | AF204265.1 | - | 6 | 634 | ||
AF204266.1 | - | 5 | 595 | |||
AF204267.1 | - | 6 | 625 | |||
AF204268.1 | - | 5 | 586 | |||
AF307947.1 | - | 5 | 647 | |||
AF307948.1 | - | 5 | 643 |
Gene Symbol: Klra15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013793.2 | NP_038821.2 | 5 | 577 | ||
GenBank mRNA | AF146571.2 | - | 5 | 577 | ||
AY003920.1 | - | 6 | 659 | |||
BC125466.1 | - | 5 | 629 | 40129981 | ||
BC125494.1 | - | 5 | 629 | 40130009 |
Gene Symbol: Klra20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053150.2 | NP_444380.1 | 5 | 633 | ||
GenBank mRNA | AF288379.1 | - | 5 | 633 |