Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam46b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175307.6 | NP_780516.1 | 2 | 692 | ||
GenBank mRNA | AK028944.1 | - | 2 | 689 | ||
AK135706.1 | - | 2 | 687 | |||
AK137659.1 | - | 2 | 693 | |||
AK162145.1 | - | 2 | 400 | |||
AK163215.1 | - | 2 | 692 | |||
BC038001.1 | - | 2 | 650 | 4913445 |