Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016892.3 | NP_058588.1 | 4 | 475 | ||
GenBank mRNA | AF121906.1 | - | 4 | 388 | ||
AF173379.1 | - | 4 | 448 | |||
AK010264.1 | - | 1 | 84 | |||
AK050072.1 | - | 4 | 424 | |||
AK192205.1 | - | 2 | 170 | |||
BC026938.1 | - | 4 | 442 | 4024140 |