Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010194.2 | NP_034324.2 | 3, 4 | 515 | ||
GenBank mRNA | AK004587.1 | - | 3, 4 | 507 | ||
AK143639.1 | - | 3, 4 | 465 | |||
AK155861.1 | - | 3, 4 | 511 | |||
AK170418.1 | - | 3, 4 | 456 | |||
BC038130.1 | - | 3, 4 | 529 | |||
BC100306.1 | - | 3, 4 | 440 | |||
BC129919.1 | - | 2, 3 | 451 | 40111855 | ||
X12616.1 | - | 2, 3 | 455 |