Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013536.2 | NP_038564.1 | 6 | 830 | ||
GenBank mRNA | AA185226.1 | - | 2 | 160 | ||
AK081757.1 | - | 6 | 808 | |||
AK178640.1 | - | 2 | 210 | |||
AK218607.1 | - | 4 | 351 | |||
BC002004.1 | - | 6 | 750 | 3487480 |
Gene Symbol: Lpcat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145130.2 | NP_660112.1 | 13 | 2367 | ||
GenBank mRNA | BB666850.1 | - | 1 | 497 |