Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077353.2 | NP_001070821.1 | 6 | 501 | ||
NM_001288617.1 | NP_001275546.1 | 6 | 546 | |||
NM_010356.4 | NP_034486.2 | 5 | 406 | |||
GenBank mRNA | AI047326.1 | - | 6 | 479 | ||
AI255699.1 | - | 5 | 402 | |||
AK002478.1 | - | 4 | 354 | |||
AK014076.1 | - | 5 | 379 | |||
AK149543.1 | - | 5 | 371 | |||
BC147272.1 | - | 5 | 369 | 9056327 | ||
BC147273.1 | - | 5 | 369 | 9056339 | ||
GQ227791.1 | - | 4 | 331 | |||
M73483.1 | - | 4 | 337 | |||
X65021.1 | - | 4 | 322 |