Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010464.2 | NP_034594.1 | 1 | 100 | ||
GenBank mRNA | AF193796.1 | - | 1 | 100 | ||
AY349616.1 | - | 1 | 128 |