Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myoz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF480442.1 | - | 6 | 551 | ||
BC104046.2 | - | 6 | 473 | 40040057 | ||
BC104047.1 | - | 6 | 473 | 40040058 |