Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010809.1 | NP_034939.1 | 10 | 1441 | ||
GenBank mRNA | AK148467.1 | - | 10 | 1419 | ||
BC006725.1 | - | 10 | 1405 | 3962288 | ||
X63162.1 | - | 10 | 1441 | |||
X66402.1 | - | 10 | 1383 |