Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mug2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008646.3 | NP_032672.2 | 16 | 2025 | ||
GenBank mRNA | M65238.1 | - | 16 | 2028 |