Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Proc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042767.2 | NP_001036232.1 | 7 | 724 | ||
NM_001042768.2 | NP_001036233.2 | 8 | 804 | |||
NM_008934.3 | NP_032960.3 | 7 | 736 | |||
GenBank mRNA | AF318182.1 | - | 7 | 713 | ||
BC013896.1 | - | 7 | 741 | 4211329 | ||
CA468822.1 | - | 7 | 748 | |||
CF550816.1 | - | 3 | 300 | |||
D10445.1 | - | 6 | 652 | |||
DV662175.1 | - | 6 | 638 |