Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009126.3 | NP_033152.3 | 1 | 50 | ||
GenBank mRNA | AY144683.1 | - | 1 | 34 | ||
AY144684.1 | - | 1 | 34 | |||
BC063756.1 | - | 1 | 50 | 30288465 |