Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031199.3 | NP_112476.1 | 1, 2 | 292 | ||
GenBank mRNA | AK010785.1 | - | 1, 2 | 320 | ||
AK162680.1 | - | 1, 2 | 57 | |||
AK166321.1 | - | 1, 2 | 225 | |||
BC132185.1 | - | 1, 2 | 90 | 40130462 | ||
BC132211.1 | - | 1, 2 | 90 | 40130488 | ||
M92420.1 | - | 1, 2 | 42 | |||
U65016.1 | - | 1, 2 | 127 |