Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136240.1 | NP_001129712.1 | 8 | 1771 | ||
NM_172264.2 | NP_758468.2 | 8 | 1569 | |||
NM_175343.5 | NP_780552.1 | 8 | 1744 | |||
GenBank mRNA | AK030900.1 | - | 8 | 1745 | ||
AK042454.1 | - | 1 | 67 | |||
AK052715.1 | - | 8 | 1752 | |||
AK087570.1 | - | 8 | 1680 | |||
AK164042.1 | - | 8 | 1607 | |||
BC039186.1 | - | 8 | 1569 | 3979709 |