Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282021.1 | NP_001268950.1 | 4 | 748 | ||
NM_001282022.1 | NP_001268951.1 | 4 | 718 | |||
NM_009499.3 | NP_033525.2 | 4 | 748 | |||
GenBank mRNA | AF084548.1 | - | 4 | 715 | ||
AK140329.1 | - | 4 | 712 | |||
AK150103.1 | - | 4 | 710 | |||
AK155480.1 | - | 4 | 711 | |||
AK157021.1 | - | 4 | 712 | |||
AK170780.1 | - | 4 | 674 | |||
AK171302.1 | - | 4 | 713 | |||
AK171715.1 | - | 4 | 712 | |||
BC010223.1 | - | 4 | 450 | 3500587 | ||
BC015289.1 | - | 4 | 691 | 4240907 |