Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lemd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146075.2 | NP_666187.2 | 1, 2 | 775 | ||
GenBank mRNA | AK082131.1 | - | 1, 2 | 582 | ||
AK143224.1 | - | 1, 2 | 760 | |||
AY640307.1 | - | 1, 2 | 755 | |||
BC026588.1 | - | 1, 2 | 85 | 4972900 |