Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147221.2 | NP_671754.2 | 6 | 1787 | ||
GenBank mRNA | AF220434.1 | - | 4 | 1641 | ||
AF486579.1 | - | 6 | 1786 | |||
AK145342.1 | - | 3 | 311 | |||
BC066157.1 | - | 4 | 1222 | 30468539 |