Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301407.1 | NP_001288336.1 | 5 | 667 | ||
NM_146173.3 | NP_666285.1 | 5 | 670 | |||
GenBank mRNA | AF276890.1 | - | 5 | 669 | ||
AK004954.1 | - | 5 | 644 | |||
AK162631.1 | - | 4 | 2265 | |||
AK172176.1 | - | 5 | 648 | |||
BC024685.1 | - | 5 | 628 | 3981492 | ||
BE861472.1 | - | 3 | 279 | |||
BU054333.1 | - | 5 | 421 |