Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172279.1 | NP_758483.1 | 9 | 855 | ||
GenBank mRNA | AK122565.2 | - | 5 | 501 | ||
AK146784.1 | - | 10 | 1190 | |||
AY151083.1 | - | 9 | 855 |