Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033356.3 | NP_001028528.2 | 2 | 722 | ||
NM_001289692.1 | NP_001276621.1 | 2 | 722 | |||
GenBank mRNA | AK142898.1 | - | 2 | 330 | ||
BC150901.1 | - | 2 | 315 | 9087812 | ||
BC172055.1 | - | 2 | 315 |