Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgcz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145841.2 | NP_665840.2 | 3, 4 | 1084 | ||
GenBank mRNA | AK048960.1 | - | 3, 4 | 1048 | ||
AK136779.1 | - | 3, 4 | 1085 | |||
AY095374.1 | - | 2, 3 | 300 | |||
AY250713.1 | - | 2, 3 | 300 | |||
BC104048.1 | - | 2, 3 | 300 | 40040143 | ||
BC104049.2 | - | 2, 3 | 300 | 40040144 |