Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007953.2 | NP_031979.2 | 2 | 269 | ||
GenBank mRNA | AK019222.1 | - | 2 | 230 | ||
AK046588.1 | - | 2 | 229 | |||
AK156371.1 | - | 2 | 245 | |||
BC034165.1 | - | 2 | 212 | |||
BC039774.1 | - | 2 | 281 | |||
BC138586.1 | - | 2 | 229 | 8861606 | ||
BC171958.1 | - | 2 | 229 | 9054244 | ||
CX208655.1 | - | 2 | 188 | |||
U85259.1 | - | 2 | 236 |