Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172979.3 | NP_766567.1 | 4 | 1114 | ||
GenBank mRNA | AK028832.1 | - | 3 | 309 | ||
AK052840.1 | - | 4 | 1001 | |||
AK142590.1 | - | 4 | 1001 | |||
AY198336.1 | - | 4 | 1032 | |||
AY198337.1 | - | 4 | 999 | |||
BC048853.1 | - | 1 | 43 | |||
BC055469.1 | - | 3 | 282 | |||
BC116796.1 | - | 4 | 993 | 40126115 | ||
BC119176.1 | - | 4 | 993 | 8733925 |