Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ust | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177387.3 | NP_796361.2 | 1, 2 | 675 | ||
GenBank mRNA | AK086181.1 | - | 1, 2 | 622 | ||
AK147850.1 | - | 1, 2 | 676 | |||
AK162950.1 | - | 1, 2 | 248 | |||
BC138155.1 | - | 1, 2 | 415 | 8861174 |