Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ramp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168392.1 | NP_001161864.1 | 2 | 405 | ||
NM_016894.3 | NP_058590.1 | 2 | 405 | |||
NM_178401.3 | NP_848488.2 | 2 | 405 | |||
GenBank mRNA | AB019046.1 | - | 2 | 216 | ||
AF146522.1 | - | 2 | 197 | |||
AF209904.1 | - | 2 | 176 | |||
AJ250489.1 | - | 2 | 197 | |||
AK144331.1 | - | 2 | 405 | |||
AK157896.1 | - | 2 | 195 | |||
AK165238.1 | - | 2 | 181 | |||
BC012644.1 | - | 2 | 162 | 4207524 | ||
BY707433.1 | - | 2 | 186 | |||
BY733497.1 | - | 2 | 405 | |||
CB522711.1 | - | 1 | 42 |