Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019991.1 | NP_064375.1 | 3 | 289 | ||
GenBank mRNA | AB032428.1 | - | 3 | 310 | ||
AF226610.1 | - | 3 | 245 | |||
AF234636.1 | - | 3 | 289 | |||
AF272368.1 | - | 3 | 241 | |||
AK005424.1 | - | 3 | 266 | |||
BC099458.1 | - | 3 | 285 | 30790018 |