Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itm2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022417.3 | NP_071862.2 | 3 | 533 | ||
GenBank mRNA | AB030199.1 | - | 3 | 476 | ||
AF272044.1 | - | 3 | 440 | |||
AF282981.1 | - | 3 | 461 | |||
AK036377.1 | - | 3 | 479 | |||
AK156930.1 | - | 3 | 436 | |||
BC012952.1 | - | 3 | 411 | 2648812 |