Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ostc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025509.3 | NP_079785.1 | 3 | 357 | ||
GenBank mRNA | AK009237.1 | - | 3 | 332 | ||
AK011085.1 | - | 3 | 309 | |||
AK013011.1 | - | 3 | 316 | |||
AK135332.1 | - | 3 | 313 | |||
BC014799.1 | - | 3 | 332 | |||
BC021935.1 | - | 3 | 331 | 5149713 | ||
BY228396.1 | - | 3 | 359 |