Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025881.3 | NP_080157.1 | 2, 3 | 282 | ||
NM_028190.3 | NP_082466.2 | 2, 3 | 282 | |||
GenBank mRNA | AK007781.1 | - | 3, 4 | 306 | ||
AK010539.1 | - | 2, 3 | 234 | |||
AK017655.1 | - | 2, 3 | 242 | |||
AK160293.1 | - | 2, 3 | 283 | |||
AK211801.1 | - | 3, 4 | 210 | |||
BC034135.1 | - | 1, 2 | 448 | 5003664 | ||
BC055875.1 | - | 2, 3 | 1265 | |||
BY726224.1 | - | 3, 4 | 340 |