Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026347.3 | NP_080623.2 | 6 | 776 | ||
GenBank mRNA | AI837665.1 | - | 1 | 162 | ||
AK005287.1 | - | 6 | 777 | |||
BC060949.1 | - | 6 | 779 | 6533986 | ||
BC087901.1 | - | 6 | 747 | 30303437 |