Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mum1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023431.5 | NP_075920.4 | 5 | 496 | ||
GenBank mRNA | AK078792.1 | - | 2 | 203 | ||
AK154610.1 | - | 5 | 415 | |||
AK158750.1 | - | 5 | 496 | |||
AK159972.1 | - | 4 | 1398 | |||
AK170459.1 | - | 5 | 415 | |||
AK180155.1 | - | 4 | 358 | |||
AK205449.1 | - | 4 | 357 | |||
AK210750.1 | - | 1 | 60 | |||
AK219009.1 | - | 4 | 358 | |||
AK220121.1 | - | 4 | 358 | |||
BC008550.1 | - | 2 | 172 | |||
BC075617.1 | - | 4 | 385 | 5718202 |