Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrn4cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013019.2 | NP_001013037.1 | 2 | 982 | ||
GenBank mRNA | AK087615.1 | - | 984 | |||
AK158268.1 | - | 2 | 920 | |||
BC107244.2 | - | 2 | 955 | 40052213 | ||
BC107245.2 | - | 2 | 955 | 40052216 |