Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133731.2 | NP_598492.2 | 3, 4 | 368 | ||
GenBank mRNA | AB010778.1 | - | 3, 4 | 351 | ||
AK014645.1 | - | 3, 4 | 368 | |||
BC055854.1 | - | 3, 4 | 353 | 4035351 | ||
CD773447.1 | - | 3, 4 | 372 |