Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osgin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027950.1 | NP_082226.1 | 5 | 641 | ||
GenBank mRNA | AB221676.1 | - | 4 | 468 | ||
AK005892.1 | - | 3 | 404 | |||
BC006032.1 | - | 4 | 982 | |||
BC022135.1 | - | 5 | 641 | 5099665 | ||
BC110674.1 | - | 5 | 628 | |||
BC132440.1 | - | 5 | 610 | 40130717 | ||
BC138192.1 | - | 5 | 610 | 8861211 |