Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbed3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011319.1 | - | 2 | 177 | ||
AK135692.1 | - | 2 | 238 | |||
AK135762.1 | - | 2 | 237 | |||
AK135797.1 | - | 2 | 236 | |||
AK139537.1 | - | 2 | 232 | |||
AK162164.1 | - | 2 | 236 | |||
AK162256.1 | - | 2 | 236 | |||
AK163367.1 | - | 2 | 236 | |||
BC016263.1 | - | 2 | 113 | 4921559 | ||
BC085478.1 | - | 2 | 173 | 6808900 | ||
BU709250.1 | - | 2 | 501 | |||
BY093894.1 | - | 2 | 99 |