Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028207.3 | NP_082483.1 | 3 | 500 | ||
GenBank mRNA | AF280809.1 | - | 3 | 465 | ||
AK008734.1 | - | 3 | 481 | |||
AK079346.1 | - | 3 | 381 | |||
AK170772.1 | - | 3 | 481 | |||
BC016269.1 | - | 3 | 508 | 4489142 |