Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033145.1 | NP_149157.1 | 4 | 394 | ||
GenBank mRNA | AK210530.1 | - | 1 | 61 | ||
BC048433.1 | - | 4 | 392 | 6772515 |