Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim30b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040770.1 | - | 1659 | |||
AK080092.1 | - | 1660 | ||||
AK172348.1 | - | 1557 |