Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089194.1 | - | 1481 | |||
AK129468.1 | - | 1025 | ||||
AK154663.1 | - | 1 | 691 | |||
AK158313.1 | - | 3 | 1386 | |||
AK159547.1 | - | 1558 | ||||
AK170121.1 | - | 1 | 691 | |||
AK170885.1 | - | 1447 | ||||
BC092227.1 | - | 2 | 1336 | 30531788 |